Assay Details
| Gene Symbol: Acvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277258.1 | NP_001264187.1 | 3 - 4 | 273 | 120 | |
| NM_009612.3 | NP_033742.2 | 3 - 4 | 342 | 189 | ||
| GenBank mRNA | BC015083.1 | - | 2 - 3 | 223 | 3980151 | 120 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC015083.1, NM_001277258.1, NM_009612.3 transcripts.
Set Membership: |
|

Acvrl1
Genomic Map