Assay Details
| Gene Symbol: Tead3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098226.3 | NP_001091696.2 | 7 - 8 | 615 | 53 | |
| NM_001204156.1 | NP_001191085.1 | 7 - 8 | 938 | 53 | ||
| NM_011566.4 | NP_035696.3 | 7 - 8 | 779 | 53 | ||
| GenBank mRNA | AF002670.1 | - | 7 - 8 | 734 | 53 | |
| AK087649.1 | - | 7 - 8 | 616 | 53 | ||
| AK143551.1 | - | 6 - 7 | 1651 | 53 | ||
| BC021364.1 | - | 7 - 8 | 659 | 53 | ||
| BC047929.1 | - | 7 - 8 | 938 | 53 | ||
| D87963.1 | - | 6 - 7 | 449 | 53 | ||
| D87964.1 | - | 7 - 8 | 760 | 53 | ||
| Y10027.1 | - | 6 - 7 | 541 | 53 | ||
| Y16611.1 | - | 6 - 7 | 532 | 53 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tead3
Genomic Map