Assay Details
| Gene Symbol: Stard3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021547.3 | NP_067522.1 | 11 - 12 | 1144 | 96 | |
| GenBank mRNA | AK041538.1 | - | 11 - 12 | 1100 | 96 | |
| AK150166.1 | - | 11 - 12 | 1088 | 96 | ||
| AK150254.1 | - | 11 - 12 | 1088 | 96 | ||
| AK155124.1 | - | 11 - 12 | 1083 | 96 | ||
| AK160480.1 | - | 11 - 12 | 1064 | 96 | ||
| AK197418.1 | - | 3 - 4 | 243 | 96 | ||
| BC003313.1 | - | 11 - 12 | 1061 | 3155179 | 96 | |
| BC058727.1 | - | 12 - 13 | 1231 | 96 | ||
| X82457.1 | - | 11 - 12 | 1128 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stard3
Genomic Map