Assay Details
| Gene Symbol: Naga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008669.4 | NP_032695.3 | 1 - 2 | 69 | 110 | |
| GenBank mRNA | CJ093426.1 | - | 1 - 2 | 72 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_008669.4 transcripts.
Set Membership: |
|

Naga
Genomic Map