Assay Details
| Gene Symbol: Naga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008669.4 | NP_032695.3 | 3 - 4 | 293 | 58 | |
| GenBank mRNA | AF079458.1 | - | 3 - 4 | 251 | 58 | |
| AK077116.1 | - | 3 - 4 | 265 | 58 | ||
| AK133576.1 | - | 3 - 4 | 251 | 58 | ||
| AK153002.1 | - | 3 - 4 | 279 | 58 | ||
| BC021631.1 | - | 3 - 4 | 273 | 4019197 | 58 | |
| CJ093426.1 | - | 3 - 4 | 296 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Naga
Genomic Map