Assay Details
| Gene Symbol: Pacsin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159509.1 | NP_001152981.1 | 3 - 4 | 436 | 93 | |
| NM_001159510.1 | NP_001152982.1 | 3 - 4 | 387 | 93 | ||
| NM_011862.3 | NP_035992.1 | 3 - 4 | 475 | 93 | ||
| GenBank mRNA | AF128535.1 | - | 3 - 4 | 465 | 93 | |
| AK143828.1 | - | 3 - 4 | 407 | 93 | ||
| AK154785.1 | - | 3 - 4 | 362 | 93 | ||
| AK170299.1 | - | 3 - 4 | 434 | 93 | ||
| BB860324.1 | - | 3 - 4 | 378 | 93 | ||
| BC023502.1 | - | 4 - 5 | 341 | 5031043 | 93 | |
| CA560112.1 | - | 1 - 2 | 120 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pacsin2
Genomic Map