Assay Details
| Gene Symbol: Pacsin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159509.1 | NP_001152981.1 | 7 - 8 | 1124 | 94 | |
| NM_001159510.1 | NP_001152982.1 | 7 - 8 | 1075 | 94 | ||
| NM_011862.3 | NP_035992.1 | 7 - 8 | 1163 | 94 | ||
| GenBank mRNA | AF128535.1 | - | 7 - 8 | 1153 | 94 | |
| AK154785.1 | - | 7 - 8 | 1050 | 94 | ||
| AK170299.1 | - | 7 - 8 | 1122 | 94 | ||
| BC023502.1 | - | 8 - 9 | 1029 | 5031043 | 94 | |
| CF168957.1 | - | 2 - 3 | 268 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pacsin2
Genomic Map