Assay Details
| Gene Symbol: Pacsin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159509.1 | NP_001152981.1 | 9 - 10 | 1374 | 109 | |
| NM_001159510.1 | NP_001152982.1 | 9 - 10 | 1325 | 109 | ||
| NM_011862.3 | NP_035992.1 | 9 - 10 | 1413 | 109 | ||
| GenBank mRNA | AF128535.1 | - | 9 - 10 | 1403 | 109 | |
| AK083414.1 | - | 1 - 2 | 63 | 109 | ||
| AK154785.1 | - | 9 - 10 | 1300 | 109 | ||
| AK170299.1 | - | 9 - 10 | 1372 | 109 | ||
| AK184671.1 | - | 3 - 4 | 295 | 109 | ||
| AK202810.1 | - | 1 - 2 | 101 | 109 | ||
| BC023502.1 | - | 10 - 11 | 1279 | 5031043 | 109 | |
| CF168957.1 | - | 4 - 5 | 518 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pacsin2
Genomic Map