Assay Details
| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013871.3 | NP_038899.1 | 2 - 3 | 487 | 67 | |
| GenBank mRNA | AK011286.1 | - | 1 - 2 | 527 | 67 | |
| AK165741.1 | - | 2 - 3 | 488 | 67 | ||
| BC021640.1 | - | 2 - 3 | 405 | 3979667 | 67 | |
| BY733534.1 | - | 2 - 3 | 488 | 67 | ||
| Y13439.1 | - | 2 - 3 | 423 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk12
Genomic Map