Assay Details
| Gene Symbol: Sbf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081030.2 | NP_001074499.2 | 36 - 37 | 5266 | 64 | |
| NM_001170561.1 | NP_001164032.1 | 37 - 38 | 5344 | 64 | ||
| GenBank mRNA | AK020972.1 | - | 7 - 8 | 907 | 64 | |
| AK076039.1 | - | 4 - 5 | 511 | 64 | ||
| AK129485.1 | - | 36 - 37 | 5210 | 64 | ||
| AK147332.1 | - | 36 - 37 | 5223 | 64 | ||
| BC029156.1 | - | 7 - 8 | 902 | 64 | ||
| BC098209.1 | - | 17 - 18 | 2609 | 64 | ||
| BC157935.1 | - | 36 - 37 | 5156 | 9088115 | 64 | |
| BC158013.1 | - | 37 - 38 | 5234 | 9007320 | 64 | |
| BC172094.1 | - | 36 - 37 | 5156 | 9054380 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sbf1
Genomic Map