Assay Details
Gene Symbol: Sult4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013873.3 | NP_038901.3 | 2 - 3 | 461 | 82 | |
GenBank mRNA | AF059257.1 | - | 2 - 3 | 337 | 82 | |
AK003034.1 | - | 2 - 3 | 322 | 82 | ||
AK010293.1 | - | 2 - 3 | 317 | 82 | ||
AK159034.1 | - | 2 - 3 | 353 | 82 | ||
BC051132.1 | - | 2 - 3 | 334 | 6531493 | 82 | |
BC054757.1 | - | 2 - 3 | 461 | 6413084 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
