Assay Details
| Gene Symbol: Senp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144851.5 | NP_659100.1 | 10 - 11 | 1358 | 74 | |
| GenBank mRNA | AK028565.1 | - | 10 - 11 | 1100 | 74 | |
| AK028762.1 | - | 2 - 3 | 2332 | 74 | ||
| AK035581.1 | - | 10 - 11 | 1194 | 74 | ||
| AK035723.1 | - | 11 - 12 | 1541 | 74 | ||
| AK054521.1 | - | 11 - 12 | 1490 | 74 | ||
| AK088597.1 | - | 10 - 11 | 74 | |||
| BC023129.1 | - | 9 - 10 | 1072 | 3595534 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Senp1
Genomic Map