Assay Details
| Gene Symbol: Senp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144851.5 | NP_659100.1 | 11 - 12 | 1438 | 70 | |
| GenBank mRNA | AK028565.1 | - | 11 - 12 | 1180 | 70 | |
| AK028762.1 | - | 3 - 4 | 2412 | 70 | ||
| AK035581.1 | - | 11 - 12 | 1274 | 70 | ||
| AK035723.1 | - | 12 - 13 | 1621 | 70 | ||
| AK054521.1 | - | 12 - 13 | 1570 | 70 | ||
| AK088597.1 | - | 11 - 12 | 1182 | 70 | ||
| BC023129.1 | - | 10 - 11 | 1152 | 3595534 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Senp1
Genomic Map