Assay Details
| Gene Symbol: Slc38a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027052.3 | NP_081328.2 | 12 - 13 | 1296 | 56 | |
| GenBank mRNA | AB055004.1 | - | 12 - 13 | 1272 | 56 | |
| AK051023.1 | - | 12 - 13 | 1297 | 56 | ||
| AK164292.1 | - | 12 - 13 | 1239 | 56 | ||
| AK167539.1 | - | 12 - 13 | 1278 | 56 | ||
| AK169349.1 | - | 11 - 12 | 1285 | 56 | ||
| AY027919.1 | - | 12 - 13 | 1335 | 56 | ||
| BC024072.1 | - | 12 - 13 | 1221 | 5097293 | 56 | |
| BC024123.1 | - | 12 - 13 | 1253 | 5098067 | 56 | |
| BC031717.1 | - | 12 - 13 | 1214 | 4195569 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a4
Genomic Map