Assay Details
| Gene Symbol: Slc38a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027052.3 | NP_081328.2 | 8 - 9 | 778 | 64 | |
| GenBank mRNA | AB055004.1 | - | 8 - 9 | 754 | 64 | |
| AK051023.1 | - | 8 - 9 | 779 | 64 | ||
| AK164292.1 | - | 8 - 9 | 721 | 64 | ||
| AK167539.1 | - | 8 - 9 | 760 | 64 | ||
| AK169349.1 | - | 7 - 8 | 767 | 64 | ||
| AY027919.1 | - | 8 - 9 | 817 | 64 | ||
| BC024072.1 | - | 8 - 9 | 703 | 5097293 | 64 | |
| BC024123.1 | - | 8 - 9 | 735 | 5098067 | 64 | |
| BC031717.1 | - | 8 - 9 | 696 | 4195569 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a4
Genomic Map