Assay Details
| Gene Symbol: Gm6961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038648.1 | - | 2 - 3 | 236 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | developing brain homeobox 2 | Dbx2 |
9430056A22Rik, 9630005K15, AI846217, Gm1229 |
-92.5kb | Sense | Chr. 15 - 95623563 - 95654771 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA A130051J0 gene | A130051J06Rik | ENSMUSG00000053098 | 39.1kb | Sense | Chr. 15 - 95787889 - 95805975 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm6961
Genomic Map