Assay Details
| Gene Symbol: Arid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175251.4 | NP_780460.3 | 10 - 11 | 1504 | 61 | |
| GenBank mRNA | CA749551.1 | - | 2 - 3 | 253 | 61 | |
More Information
Set Membership: |
|

Arid2
Genomic Map