Assay Details
| Gene Symbol: Prr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146061.4 | NP_666173.4 | 6 - 7 | 743 | 59 | |
| GenBank mRNA | AF482997.1 | - | 6 - 7 | 738 | 59 | |
| AK046057.1 | - | 7 - 8 | 560 | 59 | ||
| AK081231.1 | - | 6 - 7 | 739 | 59 | ||
| BC024991.1 | - | 7 - 8 | 627 | 5065129 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prr5
Genomic Map