Assay Details
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK054432.1 | - | 2 - 3 | 728 | 68 | |
AK088096.1 | - | 2 - 3 | 733 | 68 | ||
AK161059.1 | - | 2 - 3 | 717 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
