Assay Details
| Gene Symbol: Rabl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026817.3 | NP_081093.2 | 1 - 2 | 106 | 81 | |
| GenBank mRNA | AK051232.1 | - | 1 - 2 | 89 | 81 | |
| AK170205.1 | - | 1 - 2 | 49 | 81 | ||
| BC030376.1 | - | 1 - 2 | 73 | 5374611 | 81 | |
| BQ887131.1 | - | 1 - 2 | 98 | 81 | ||
| BY273527.1 | - | 1 - 2 | 106 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK170205.1, BC030376.1, NM_026817.3 transcripts.
Set Membership: |
|

Rabl2
Genomic Map