Assay Details
| Gene Symbol: Rabl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026817.3 | NP_081093.2 | 3 - 4 | 291 | 78 | |
| GenBank mRNA | AK004012.1 | - | 3 - 4 | 237 | 78 | |
| AK051232.1 | - | 3 - 4 | 274 | 78 | ||
| BC030376.1 | - | 3 - 4 | 258 | 5374611 | 78 | |
| BQ887131.1 | - | 3 - 4 | 283 | 78 | ||
| BY273527.1 | - | 3 - 4 | 291 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabl2
Genomic Map