Assay Details
| Gene Symbol: Alg12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142357.1 | NP_001135829.1 | 7 - 8 | 1071 | 90 | |
| GenBank mRNA | AJ429133.1 | - | 7 - 8 | 1031 | 90 | |
| AK142322.1 | - | 7 - 8 | 1072 | 90 | ||
| AK179946.1 | - | 2 - 3 | 272 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alg12
Genomic Map