Assay Details
| Gene Symbol: L3mbtl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289711.1 | NP_001276640.1 | 13 - 14 | 1703 | 97 | |
| NM_001289712.1 | NP_001276641.1 | 13 - 14 | 1703 | 97 | ||
| NM_145993.5 | NP_666105.2 | 13 - 14 | 1703 | 97 | ||
| NR_110362.1 | 13 - 14 | 2026 | 97 | |||
| GenBank mRNA | AK029115.1 | - | 13 - 14 | 1648 | 97 | |
| AK036510.1 | - | 13 - 14 | 1656 | 97 | ||
| AK132012.1 | - | 13 - 14 | 1655 | 97 | ||
| AK141810.1 | - | 12 - 13 | 2836 | 97 | ||
| AK145560.1 | - | 13 - 14 | 1645 | 97 | ||
| AK146632.1 | - | 13 - 14 | 1659 | 97 | ||
| AK156236.1 | - | 13 - 14 | 1658 | 97 | ||
| AK211064.1 | - | 3 - 4 | 341 | 97 | ||
| AY237000.1 | - | 13 - 14 | 1636 | 97 | ||
| AY237001.1 | - | 13 - 14 | 1636 | 97 | ||
| AY237002.1 | - | 13 - 14 | 1959 | 97 | ||
| BC023676.1 | - | 2 - 3 | 238 | 97 | ||
| BC023933.1 | - | 13 - 14 | 1644 | 5325131 | 97 | |
| BC030864.1 | - | 13 - 14 | 1606 | 4211489 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

L3mbtl2
Genomic Map