Assay Details
| Gene Symbol: L3mbtl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289711.1 | NP_001276640.1 | 1 - 2 | 141 | 69 | |
| NM_001289712.1 | NP_001276641.1 | 1 - 2 | 141 | 69 | ||
| NM_145993.5 | NP_666105.2 | 1 - 2 | 141 | 69 | ||
| NR_110362.1 | 1 - 2 | 141 | 69 | |||
| GenBank mRNA | AK029115.1 | - | 1 - 2 | 86 | 69 | |
| AK036510.1 | - | 1 - 2 | 94 | 69 | ||
| AK132012.1 | - | 1 - 2 | 94 | 69 | ||
| AK141810.1 | - | 1 - 2 | 42 | 69 | ||
| AK145560.1 | - | 1 - 2 | 83 | 69 | ||
| AK146632.1 | - | 1 - 2 | 97 | 69 | ||
| AK156236.1 | - | 1 - 2 | 96 | 69 | ||
| AY237000.1 | - | 1 - 2 | 74 | 69 | ||
| AY237001.1 | - | 1 - 2 | 74 | 69 | ||
| BC023933.1 | - | 1 - 2 | 82 | 5325131 | 69 | |
| BC030864.1 | - | 1 - 2 | 44 | 4211489 | 69 | |
| BY130665.1 | - | 1 - 2 | 142 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029115.1, AK036510.1, AK145560.1, AK156236.1, AY237000.1, AY237001.1, BC023933.1, BC030864.1, NM_001289711.1, NM_001289712.1, NM_145993.5 transcripts.
Set Membership: |
|

L3mbtl2
Genomic Map