Assay Details
| Gene Symbol: Xpnpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177310.2 | NP_796284.1 | 5 - 6 | 893 | 85 | |
| GenBank mRNA | AK088324.1 | - | 5 - 6 | 893 | 85 | |
| BC137569.1 | - | 5 - 6 | 877 | 8860588 | 85 | |
| BC144717.1 | - | 5 - 6 | 878 | 9053265 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xpnpep3
Genomic Map