Assay Details
| Gene Symbol: Xpnpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC137569.1 | - | 7 - 8 | 1074 | 8860588 | 89 |
| BC144717.1 | - | 7 - 8 | 1075 | 9053265 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xpnpep3
Genomic Map