Assay Details
| Gene Symbol: Pmm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013872.4 | NP_038900.1 | 3 - 4 | 407 | 111 | |
| GenBank mRNA | AF007267.1 | - | 3 - 4 | 278 | 111 | |
| AK004631.1 | - | 3 - 4 | 340 | 111 | ||
| AK013805.1 | - | 3 - 4 | 311 | 111 | ||
| AK166999.1 | - | 3 - 4 | 342 | 111 | ||
| CX217041.1 | - | 3 - 4 | 407 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pmm1
Genomic Map