Assay Details
| Gene Symbol: Dhx40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026191.2 | NP_080467.3 | 2 - 3 | 391 | 113 | |
| GenBank mRNA | AK004610.1 | - | 2 - 3 | 355 | 113 | |
| AK010512.1 | - | 2 - 3 | 371 | 113 | ||
| AK029935.1 | - | 2 - 3 | 364 | 113 | ||
| AK031000.1 | - | 2 - 3 | 338 | 113 | ||
| AK135592.1 | - | 2 - 3 | 327 | 113 | ||
| BC058276.1 | - | 2 - 3 | 309 | 6515479 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004610.1, AK010512.1, AK031000.1, BC058276.1 transcripts.
Set Membership: |
|

Dhx40
Genomic Map