Assay Details
| Gene Symbol: Tmprss12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183109.3 | NP_898932.2 | 1 - 2 | 192 | 71 | |
| GenBank mRNA | AK015583.1 | - | 1 - 2 | 205 | 71 | |
| AK132967.1 | - | 1 - 2 | 193 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015583.1 transcripts.
Set Membership: |
|

Tmprss12
Genomic Map