Assay Details
| Gene Symbol: Fyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011815.5 | NP_035945.1 | 10 - 11 | 1979 | 81 | |
| GenBank mRNA | AK172164.1 | - | 3 - 4 | 211 | 81 | |
| BC141352.1 | - | 10 - 11 | 1920 | 9055964 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fyb
Genomic Map