Assay Details
| Gene Symbol: Fyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278269.1 | NP_001265198.1 | 13 - 14 | 2169 | 124 | |
| NM_011815.5 | NP_035945.1 | 14 - 15 | 2307 | 124 | ||
| GenBank mRNA | AF001863.1 | - | 13 - 14 | 2145 | 124 | |
| AF061744.1 | - | 14 - 15 | 2283 | 124 | ||
| AK038225.1 | - | 13 - 14 | 2197 | 124 | ||
| AK137907.1 | - | 13 - 14 | 2125 | 124 | ||
| AK172164.1 | - | 7 - 8 | 539 | 124 | ||
| BC141352.1 | - | 14 - 15 | 2248 | 9055964 | 124 | |
More Information
Set Membership: |
|

Fyb
Genomic Map