Assay Details
| Gene Symbol: Fyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278269.1 | NP_001265198.1 | 1 - 2 | 1202 | 68 | |
| NM_011815.5 | NP_035945.1 | 1 - 2 | 1202 | 68 | ||
| GenBank mRNA | AF001863.1 | - | 1 - 2 | 1178 | 68 | |
| AF061744.1 | - | 1 - 2 | 1178 | 68 | ||
| AK037454.1 | - | 1 - 2 | 1203 | 68 | ||
| AK038225.1 | - | 1 - 2 | 1186 | 68 | ||
| AK137907.1 | - | 1 - 2 | 1157 | 68 | ||
| AK172013.1 | - | 1 - 2 | 1158 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fyb
Genomic Map