Assay Details
| Gene Symbol: Scpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029023.3 | NP_083299.3 | 4 - 5 | 540 | 89 | |
| GenBank mRNA | AF330052.1 | - | 4 - 5 | 496 | 89 | |
| AK014680.1 | - | 4 - 5 | 541 | 89 | ||
| AK050181.1 | - | 4 - 5 | 478 | 89 | ||
| AK153225.1 | - | 4 - 5 | 528 | 89 | ||
| AK186804.1 | - | 1 - 2 | 54 | 89 | ||
| AK188859.1 | - | 1 - 2 | 51 | 89 | ||
| AK206268.1 | - | 2 - 3 | 235 | 89 | ||
| BC004847.1 | - | 4 - 5 | 530 | 2650587 | 89 | |
| BC021399.1 | - | 4 - 5 | 495 | 4193674 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Scpep1
Genomic Map