Assay Details
| Gene Symbol: Scpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029023.3 | NP_083299.3 | 5 - 6 | 614 | 60 | |
| GenBank mRNA | AF330052.1 | - | 5 - 6 | 570 | 60 | |
| AK014680.1 | - | 5 - 6 | 615 | 60 | ||
| AK050181.1 | - | 5 - 6 | 552 | 60 | ||
| AK153225.1 | - | 5 - 6 | 602 | 60 | ||
| AK153485.1 | - | 4 - 5 | 403 | 60 | ||
| AK188859.1 | - | 2 - 3 | 125 | 60 | ||
| BC004847.1 | - | 5 - 6 | 604 | 2650587 | 60 | |
| BC021399.1 | - | 5 - 6 | 569 | 4193674 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scpep1
Genomic Map