Assay Details
| Gene Symbol: Agxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031851.1 | NP_001027021.1 | 2 - 3 | 193 | 83 | |
| GenBank mRNA | AK149538.1 | - | 2 - 3 | 204 | 83 | |
| BC141248.1 | - | 2 - 3 | 217 | 9055860 | 83 | |
| BC145576.1 | - | 2 - 3 | 214 | 9054142 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agxt2
Genomic Map