Assay Details
| Gene Symbol: Agxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031851.1 | NP_001027021.1 | 4 - 5 | 501 | 64 | |
| GenBank mRNA | AK149538.1 | - | 4 - 5 | 512 | 64 | |
| BC141248.1 | - | 4 - 5 | 525 | 9055860 | 64 | |
| BC145576.1 | - | 4 - 5 | 522 | 9054142 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agxt2
Genomic Map