Assay Details
| Gene Symbol: Agxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031851.1 | NP_001027021.1 | 6 - 7 | 691 | 88 | |
| GenBank mRNA | AK149538.1 | - | 6 - 7 | 702 | 88 | |
| BC141248.1 | - | 6 - 7 | 715 | 9055860 | 88 | |
| BC145576.1 | - | 6 - 7 | 712 | 9054142 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agxt2
Genomic Map