Assay Details
| Gene Symbol: Gsdma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029727.2 | NP_084003.2 | 2 - 3 | 131 | 68 | |
| GenBank mRNA | AK008613.1 | - | 2 - 3 | 128 | 68 | |
| AK008663.1 | - | 2 - 3 | 128 | 68 | ||
| BC109336.1 | - | 2 - 3 | 65 | 40052146 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008613.1, BC109336.1, NM_029727.2 transcripts.
Set Membership: |
|

Gsdma2
Genomic Map