Assay Details
| Gene Symbol: Adap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172133.1 | NP_742145.1 | 4 - 5 | 452 | 74 | |
| GenBank mRNA | AK050079.1 | - | 4 - 5 | 510 | 74 | |
| AK050398.1 | - | 4 - 5 | 501 | 74 | ||
| AK053152.1 | - | 4 - 5 | 496 | 74 | ||
| AK139005.1 | - | 4 - 5 | 504 | 74 | ||
| AK140864.1 | - | 4 - 5 | 483 | 74 | ||
| BC027165.1 | - | 4 - 5 | 452 | 4951680 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adap2
Genomic Map