Assay Details
| Gene Symbol: Adap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172133.1 | NP_742145.1 | 5 - 6 | 561 | 76 | |
| GenBank mRNA | AK050079.1 | - | 5 - 6 | 619 | 76 | |
| AK050398.1 | - | 5 - 6 | 610 | 76 | ||
| AK053152.1 | - | 5 - 6 | 605 | 76 | ||
| AK139005.1 | - | 5 - 6 | 613 | 76 | ||
| AK140864.1 | - | 5 - 6 | 592 | 76 | ||
| BC027165.1 | - | 5 - 6 | 561 | 4951680 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adap2
Genomic Map