Assay Details
| Gene Symbol: Msi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054043.3 | NP_473384.1 | 6 - 7 | 538 | 142 | |
| GenBank mRNA | AK031365.1 | - | 6 - 7 | 411 | 142 | |
| AK049637.1 | - | 6 - 7 | 538 | 142 | ||
| AK049688.1 | - | 6 - 7 | 538 | 142 | ||
| AK051269.1 | - | 6 - 7 | 388 | 142 | ||
| AK153928.1 | - | 6 - 7 | 411 | 142 | ||
| AK169846.1 | - | 6 - 7 | 411 | 142 | ||
| BC111809.1 | - | 7 - 8 | 471 | 40045350 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Msi2
Genomic Map