Assay Details
| Gene Symbol: Mbtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134012.3 | NP_598773.2 | 2 - 3 | 395 | 66 | |
| GenBank mRNA | AK141155.1 | - | 2 - 3 | 391 | 66 | |
| BC062907.1 | - | 2 - 3 | 395 | 6813849 | 66 | |
| BC064014.1 | - | 4 - 5 | 499 | 6335954 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mbtd1
Genomic Map