Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100032921.mm.1 |
NM_001195003.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032921.mm.1 |
NM_001195003.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032921.mm.1 |
NM_001195004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032921.mm.1 |
NM_001195004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032921.mm.1 |
TC1100003588.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100032919.mm.1 |
NM_001195003.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032919.mm.1 |
NM_001195003.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032919.mm.1 |
NM_001195004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032919.mm.1 |
NM_001195004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032919.mm.1 |
NM_177619.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032919.mm.1 |
NM_177619.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032919.mm.1 |
TC1100003588.mm.1 |
MTA TC |