| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100013809.mm.1 |
NM_008270.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013809.mm.1 |
NM_008270.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013809.mm.1 |
TC1100001492.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100013811.mm.1 |
NM_008270.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013811.mm.1 |
NM_008270.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013811.mm.1 |
TC1100001492.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100013812.mm.1 |
NM_008270.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013812.mm.1 |
NM_008270.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013812.mm.1 |
TC1100001492.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100013810.mm.1 |
NM_008270.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013810.mm.1 |
NM_008270.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013810.mm.1 |
TC1100001492.mm.1 |
MTA TC |