| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200013704.mm.1 |
NM_134156.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013704.mm.1 |
NM_134156.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013704.mm.1 |
TC1200001990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013706.mm.1 |
NM_134156.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013706.mm.1 |
NM_134156.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013706.mm.1 |
TC1200001990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013703.mm.1 |
NM_134156.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013703.mm.1 |
NM_134156.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013703.mm.1 |
TC1200001990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013705.mm.1 |
NM_134156.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013705.mm.1 |
NM_134156.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013705.mm.1 |
TC1200001990.mm.1 |
MTA TC |