Assay Details
| Gene Symbol: Actn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134156.2 | NP_598917.1 | 16 - 17 | 2230 | 68 | |
| GenBank mRNA | AK142591.1 | - | 7 - 8 | 946 | 68 | |
| AK154281.1 | - | 16 - 17 | 2223 | 68 | ||
| BC003232.1 | - | 4 - 5 | 475 | 68 | ||
| BC023206.1 | - | 3 - 4 | 432 | 68 | ||
| BC054830.1 | - | 16 - 17 | 2175 | 6308412 | 68 | |
| DQ288940.1 | - | 16 - 17 | 1950 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actn1
Genomic Map