Assay Details
| Gene Symbol: Actn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134156.2 | NP_598917.1 | 18 - 19 | 2559 | 63 | |
| GenBank mRNA | AK142591.1 | - | 9 - 10 | 1275 | 63 | |
| BC054830.1 | - | 18 - 19 | 2504 | 6308412 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Actn1
Genomic Map