| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200013719.mm.1 |
NM_134156.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013719.mm.1 |
NM_134156.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013719.mm.1 |
TC1200001990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013716.mm.1 |
NM_134156.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013716.mm.1 |
NM_134156.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013716.mm.1 |
TC1200001990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013714.mm.1 |
NM_134156.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013714.mm.1 |
NM_134156.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013714.mm.1 |
TC1200001990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013715.mm.1 |
NM_134156.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013715.mm.1 |
NM_134156.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013715.mm.1 |
TC1200001990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013718.mm.1 |
NM_134156.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013718.mm.1 |
NM_134156.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013718.mm.1 |
TC1200001990.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013717.mm.1 |
NM_134156.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013717.mm.1 |
NM_134156.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013717.mm.1 |
TC1200001990.mm.1 |
MTA TC |