Assay Details
| Gene Symbol: Actn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134156.2 | NP_598917.1 | 8 - 9 | 1046 | 102 | |
| GenBank mRNA | AK154281.1 | - | 8 - 9 | 1039 | 102 | |
| BC054830.1 | - | 8 - 9 | 991 | 6308412 | 102 | |
| DQ288940.1 | - | 8 - 9 | 766 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actn1
Genomic Map