Assay Details
| Gene Symbol: Actn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134156.2 | NP_598917.1 | 9 - 10 | 1139 | 95 | |
| GenBank mRNA | AK154281.1 | - | 9 - 10 | 1132 | 95 | |
| BC054830.1 | - | 9 - 10 | 1084 | 6308412 | 95 | |
| DQ288940.1 | - | 9 - 10 | 859 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actn1
Genomic Map